Learn More
Biomatik Corporation Native Hemoglobin (HB), 100UG
Supplier:  Biomatik Corporation RPU50307
Encompass_Preferred
Native Simian Hemoglobin (HB) is a purified Natural Protein. Purity: >90% by SDS-PAGE. Host: Simian. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Simian. Target Name: Hemoglobin (HB). Target Synonyms: Hgb; Haemoglobin; Heterotetramer(αβ)2. Accession Number: P63108; P68223. Theoretical MW: 14/16kDa. Buffer: PBS, pH7.4, containing 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5792