Learn More
Biomatik Corporation Recombinant TATA Binding Protein (TBP), 100UG
Supplier:  Biomatik Corporation RPU50712
Encompass_Preferred
Recombinant Human TATA Binding Protein (TBP) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: TATA Binding Protein (TBP). Target Synonyms: GTF2D; GTF2D1; SCA17; TFIID; TATA sequence-binding protein; TATA-box factor; Transcription initiation factor TFIID TBP subunit. Accession Number: P20226. Gene ID: 6908. Expression Region: Ile25~Thr339. Tag Info: N-terminal His Tag. Theoretical MW: 44kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5800