Learn More
Biomatik Corporation OVA Conjugated Progesterone (PG), 100UG
Catalog No. 501505803
Encompass_Preferred
OVA Conjugated Progesterone (PG) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Universal. Target Name: Progesterone (PG). Target Synonyms: P4; Pregn-4-Ene-3,20-Dione. Tag Info: N-terminal His Tag. Theoretical MW: 314.5Da. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5803 Supplier Biomatik Corporation Supplier No. RPU50786
OVA Conjugated Progesterone (PG) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Universal. Target Name: Progesterone (PG). Target Synonyms: P4; Pregn-4-Ene-3,20-Dione. Tag Info: N-terminal His Tag. Theoretical MW: 314.5Da. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.