Learn More
Biomatik Corporation OVA conjugated Pepsin (PP), 100UG
Catalog No. 501505805
Encompass_Preferred
OVA Conjugated Human Pepsin (PP) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Pepsin (PP). Accession Number: P0DJD8. Gene ID: 643834. Expression Region: Val63~Phe77. Tag Info: N-terminal His Tag. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5805 Supplier Biomatik Corporation Supplier No. RPU50805
OVA Conjugated Human Pepsin (PP) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Pepsin (PP). Accession Number: P0DJD8. Gene ID: 643834. Expression Region: Val63~Phe77. Tag Info: N-terminal His Tag. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.