Learn More
Biomatik Corporation OVA conjugated N-Telopeptide of Type I Collagen (NTXI), 100UG
Catalog No. 501505806
Encompass_Preferred
OVA Conjugated Human Cross Linked N-Telopeptide Of Type I Collagen (NTXI) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cross Linked N-Telopeptide Of Type I Collagen (NTXI). Target Synonyms: NTX-I; NTX1. Accession Number: P02452. Gene ID: 1277. Expression Region: Gln162~Pro178. Tag Info: N-terminal His Tag. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5806 Supplier Biomatik Corporation Supplier No. RPU50808
OVA Conjugated Human Cross Linked N-Telopeptide Of Type I Collagen (NTXI) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cross Linked N-Telopeptide Of Type I Collagen (NTXI). Target Synonyms: NTX-I; NTX1. Accession Number: P02452. Gene ID: 1277. Expression Region: Gln162~Pro178. Tag Info: N-terminal His Tag. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.