Learn More
Biomatik Corporation BSA conjugated C-Telopeptide of Type I Collagen (CTXI), 100UG
Supplier:  Biomatik Corporation RPU50810
Encompass_Preferred
BSA Conjugated Human Cross Linked C-Telopeptide Of Type I Collagen (CTXI) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cross Linked C-Telopeptide Of Type I Collagen (CTXI). Target Synonyms: CTX-I; CTX1. Accession Number: Q9NPZ5. Gene ID: 135152. Expression Region: Glu1207~Ala1218. Tag Info: N-terminal His Tag. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5807