Learn More
Biomatik Corporation OVA conjugated Procollagen I N-Terminal Propeptide (PINP), 100UG
Supplier:  Biomatik Corporation RPU50840
Encompass_Preferred
OVA Conjugated Human Procollagen I N-Terminal Propeptide (PINP) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Procollagen I N-Terminal Propeptide (PINP). Target Synonyms: P1NP; N-Propeptide Of Type I Procollagen; Procollagen I Amino Terminal Propeptide. Accession Number: P02452. Gene ID: 1277. Tag Info: N-terminal His Tag. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5808