Learn More
Biomatik Corporation Recombinant Human Complement C3 (C3), partial, 100UG
Supplier:  Biomatik Corporation RPC20196
Encompass_Preferred
Recombinant Human Complement C3 (C3), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: C3. Target Synonyms: C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1. Accession Number: P01024. Expression Region: 26~225aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 26.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4657