Learn More
Biomatik Corporation Recombinant Luteinizing Hormone Beta Polypeptide (LHb), 100UG
Supplier:  Biomatik Corporation RPU50994
Encompass_Preferred
Recombinant Human Luteinizing Hormone Beta Polypeptide (LHb) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Luteinizing Hormone Beta Polypeptide (LHb). Target Synonyms: LH-B; CGB4; LSH-B; hLHB; Lutropin Subunit Beta; Interstitial Cell Stimulating Hormone,Beta Chain. Accession Number: P01229. Gene ID: 3972. Expression Region: Glu23~Leu141. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5812