Learn More
Biomatik Corporation Recombinant Interleukin 10 (IL10), 100UG
Catalog No. 501505814
Encompass_Preferred
Recombinant Human Interleukin 10 (IL10) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interleukin 10 (IL10). Target Synonyms: CSIF; IL10A; TGIF; Cytokine Synthesis Inhibitory Factor. Accession Number: P22301. Gene ID: 3586. Expression Region: Met1~Asn178. Tag Info: N-terminal His Tag. Theoretical MW: 21/24/27/35kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5814 Supplier Biomatik Corporation Supplier No. RPU51130
Recombinant Human Interleukin 10 (IL10) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interleukin 10 (IL10). Target Synonyms: CSIF; IL10A; TGIF; Cytokine Synthesis Inhibitory Factor. Accession Number: P22301. Gene ID: 3586. Expression Region: Met1~Asn178. Tag Info: N-terminal His Tag. Theoretical MW: 21/24/27/35kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.