Learn More
Biomatik Corporation Recombinant Adiponectin (ADP), 100UG
Supplier:  Biomatik Corporation RPU51353
Encompass_Preferred
Recombinant Mouse Adiponectin (ADPN) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Adiponectin (ADPN). Target Synonyms: GBP28; ApM1; AdipoQ; Acrp30; ACDC; APM1; C1Q And Collagen Domain Containing; Adipocyte Complement-Related Protein Of 30 KDa; Adipose Most Abundant Gene Transcript 1. Accession Number: Q60994. Expression Region: Ala111~Asn247. Tag Info: N-terminal His and GST Tag. Theoretical MW: 46kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5817