Learn More
Biomatik Corporation Recombinant Choline Acetyltransferase (ChAT), 100UG
Catalog No. 501505818
Encompass_Preferred
Recombinant Human Choline Acetyltransferase (ChAT) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Choline Acetyltransferase (ChAT). Target Synonyms: CMS1A; CMS1A2; CHOACTase; Choline acetylase. Accession Number: P28329. Gene ID: 1103. Expression Region: Asp517~Pro732. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5818 Supplier Biomatik Corporation Supplier No. RPU51372
Recombinant Human Choline Acetyltransferase (ChAT) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Choline Acetyltransferase (ChAT). Target Synonyms: CMS1A; CMS1A2; CHOACTase; Choline acetylase. Accession Number: P28329. Gene ID: 1103. Expression Region: Asp517~Pro732. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.