Learn More
Biomatik Corporation Recombinant Human Metalloproteinase inhibitor 3 (TIMP3), partial, 100UG
Supplier:  Biomatik Corporation RPC20198
Encompass_Preferred
Recombinant Human Metalloproteinase inhibitor 3 (TIMP3), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: TIMP3. Target Synonyms: Protein MIG-5Tissue inhibitor of metalloproteinases 3; TIMP-3. Accession Number: P35625. Expression Region: 30~208aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 24.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4658