Learn More
Biomatik Corporation Recombinant N-Terminal Pro-Atrial Natriuretic Peptide (NT-ProANP), 100UG
Catalog No. 501505821
Encompass_Preferred
Recombinant Human N-Terminal Pro-Atrial Natriuretic Peptide (NT-ProANP) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: N-Terminal Pro-Atrial Natriuretic Peptide (NT-ProANP). Target Synonyms: NT-Pro-ANP; N-ANP. Accession Number: P01160. Gene ID: 4878. Expression Region: Ala25~Arg123. Tag Info: N-terminal His and GST Tag. Theoretical MW: 41kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5821 Supplier Biomatik Corporation Supplier No. RPU51490
Recombinant Human N-Terminal Pro-Atrial Natriuretic Peptide (NT-ProANP) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: N-Terminal Pro-Atrial Natriuretic Peptide (NT-ProANP). Target Synonyms: NT-Pro-ANP; N-ANP. Accession Number: P01160. Gene ID: 4878. Expression Region: Ala25~Arg123. Tag Info: N-terminal His and GST Tag. Theoretical MW: 41kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.