Learn More
Biomatik Corporation Recombinant Receptor Interacting Serine Threonine Kinase 1 (RIPK1), 100UG
Catalog No. 501505826
Encompass_Preferred
Recombinant Human Receptor Interacting Serine Threonine Kinase 1 (RIPK1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Receptor Interacting Serine Threonine Kinase 1 (RIPK1). Target Synonyms: RIP; Receptor-Interacting Protein; Cell death protein RIP; Serine/threonine-protein kinase RIP. Accession Number: Q13546. Gene ID: 8737. Expression Region: Phe17~Tyr289. Tag Info: N-terminal His Tag. Theoretical MW: 35kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5826 Supplier Biomatik Corporation Supplier No. RPU51601
Recombinant Human Receptor Interacting Serine Threonine Kinase 1 (RIPK1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Receptor Interacting Serine Threonine Kinase 1 (RIPK1). Target Synonyms: RIP; Receptor-Interacting Protein; Cell death protein RIP; Serine/threonine-protein kinase RIP. Accession Number: Q13546. Gene ID: 8737. Expression Region: Phe17~Tyr289. Tag Info: N-terminal His Tag. Theoretical MW: 35kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.