Learn More
Biomatik Corporation Recombinant Thrombopoietin (TPO), 100UG
Supplier:  Biomatik Corporation RPU51727
Encompass_Preferred
Recombinant Human Thrombopoietin (TPO) is a purified Prokaryotic Protein. Purity: >80% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Thrombopoietin (TPO). Target Synonyms: THPO; MGDF; MKCSF; ML; MPLLG; Myeloproliferative Leukemia Virus Oncogene Ligand; Megakaryocyte Growth And Development Factor; Megakaryocyte Stimulating Factor. Accession Number: P40225. Gene ID: 7066. Expression Region: Ser22~Gly349. Tag Info: N-terminal His Tag. Theoretical MW: 36kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5827