Learn More
Biomatik Corporation Recombinant Collagen Type I Alpha 2 (COL1a2), 100UG
Catalog No. 501505828
Encompass_Preferred
Recombinant Human Collagen Type I Alpha 2 (COL1a2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Collagen Type I Alpha 2 (COL1a2). Target Synonyms: COL1-A2; OI4; Collagen Alpha-2(I)chain; osteogenesis imperfecta type IV; Collagen Of Skin,Tendon And bone,Alpha-2 Chain. Accession Number: P08123. Gene ID: 1278. Expression Region: Val1103~Lys1366. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5828 Supplier Biomatik Corporation Supplier No. RPU51729
Recombinant Human Collagen Type I Alpha 2 (COL1a2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Collagen Type I Alpha 2 (COL1a2). Target Synonyms: COL1-A2; OI4; Collagen Alpha-2(I)chain; osteogenesis imperfecta type IV; Collagen Of Skin,Tendon And bone,Alpha-2 Chain. Accession Number: P08123. Gene ID: 1278. Expression Region: Val1103~Lys1366. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.