Learn More
Biomatik Corporation Recombinant Coagulation Factor XIII A1 Polypeptide (F13A1), 100UG
Supplier:  Biomatik Corporation RPU51757
Encompass_Preferred
Recombinant Human Coagulation Factor XIII A1 Polypeptide (F13A1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Coagulation Factor XIII A1 Polypeptide (F13A1). Target Synonyms: F13-A1; FXIIIA1; Protein-glutamine gamma-glutamyltransferase A chain; Transglutaminase A chain; Coagulation factor XIII A chain. Accession Number: P00488. Gene ID: 2162. Expression Region: Gly107~Asn345. Tag Info: N-terminal His Tag. Theoretical MW: 31kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5829