Learn More
Biomatik Corporation Recombinant Sirtuin 3 (SIRT3), 100UG
Catalog No. 501505833
Encompass_Preferred
Recombinant Mouse Sirtuin 3 (SIRT3) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Sirtuin 3 (SIRT3). Target Synonyms: SIR2L3; Silent Mating Type Information Regulation 2 Homolog 3; NAD-dependent protein deacetylase sirtuin-3, mitochondrial; SIR2-like protein 3; Regulatory protein SIR2 homolog 3. Accession Number: Q8R104. Gene ID: 64384. Expression Region: Gly5~Arg257. Tag Info: N-terminal His Tag. Theoretical MW: 29/16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5833 Supplier Biomatik Corporation Supplier No. RPU51870
Recombinant Mouse Sirtuin 3 (SIRT3) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Sirtuin 3 (SIRT3). Target Synonyms: SIR2L3; Silent Mating Type Information Regulation 2 Homolog 3; NAD-dependent protein deacetylase sirtuin-3, mitochondrial; SIR2-like protein 3; Regulatory protein SIR2 homolog 3. Accession Number: Q8R104. Gene ID: 64384. Expression Region: Gly5~Arg257. Tag Info: N-terminal His Tag. Theoretical MW: 29/16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.