Learn More
Biomatik Corporation Recombinant Alpha-1,4-Galactosyltransferase (a4GALT), 100UG
Supplier:  Biomatik Corporation RPU51931
Encompass_Preferred
Recombinant Human Alpha-1,4-Galactosyltransferase (a4GALT) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Alpha-1,4-Galactosyltransferase (a4GALT). Target Synonyms: A4GALT; P1; A14GALT; A4GALT1; PK; Lactosylceramide 4-Alpha-Galactosyltransferase; Globotriaosylceramide Synthase; Gb3 synthase; P1/Pk synthase; CD77 Synthase. Accession Number: Q9NPC4. Gene ID: 53947. Expression Region: Gly44~Leu353. Tag Info: N-terminal His Tag. Theoretical MW: 39kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5834