Learn More
Biomatik Corporation Recombinant Elastase 1, Pancreatic (ELA1), 100UG
Supplier:  Biomatik Corporation RPU52063
Encompass_Preferred
Recombinant Human Pancreatic Elastase 1 (ELA1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Pancreatic Elastase 1 (ELA1). Target Synonyms: PE; CELA1; Chymotrypsin-Like Elastase Family,Member 1. Accession Number: Q9UNI1. Gene ID: 1990. Expression Region: Ser40~Asn258. Tag Info: N-terminal His and GST Tag. Theoretical MW: 53kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5838