Learn More
Biomatik Corporation Recombinant Paraoxonase 3 (PON3), 100UG
Supplier:  Biomatik Corporation RPU52214
Encompass_Preferred
Recombinant Human Paraoxonase 3 (PON3) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Paraoxonase 3 (PON3). Target Synonyms: Serum paraoxonase/lactonase 3. Accession Number: Q15166. Gene ID: 5446. Expression Region: Gly2~Leu354. Tag Info: N-terminal His Tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5841