Learn More
Biomatik Corporation Recombinant TTK Protein Kinase (TTK), 100UG
Supplier:  Biomatik Corporation RPU52309
Encompass_Preferred
Recombinant Human TTK Protein Kinase (TTK) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: TTK Protein Kinase (TTK). Target Synonyms: MPS1; MPS1L1; CT96; PYT; Cancer/Testis Antigen 96; Phosphotyrosine picked threonine-protein kinase; Dual specificity protein kinase TTK. Accession Number: P33981. Gene ID: 7272. Expression Region: Gly532~Leu786. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5843