Learn More
Biomatik Corporation OVA Conjugated 25-Hydroxyvitamin D3 (HVD3), 100UG
Supplier:  Biomatik Corporation RPU52383
Encompass_Preferred
OVA Conjugated 25-Hydroxyvitamin D3 (HVD3) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Universal. Target Name: 25-Hydroxyvitamin D3 (HVD3). Target Synonyms: 25-HVD3; 25HVD3; Calcifediol; Calcidiol; H-VD3; 25-Hydroxycholecalciferol; 25-Hydroxy Vitamin D3; 25(OH)D. Tag Info: N-terminal His Tag. Theoretical MW: 400.6Da. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5844