Learn More
Biomatik Corporation Recombinant Apoptosis Inhibitor 5 (API5), 100UG
Supplier:  Biomatik Corporation RPU52476
Encompass_Preferred
Recombinant Human Apoptosis Inhibitor 5 (API5) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Apoptosis Inhibitor 5 (API5). Target Synonyms: MIG8; FIF; AAC11; API5L1; XAGL; Cell migration-inducing gene 8; Antiapoptosis clone 11; Fibroblast Growth Factor 2-Interacting Factor 2; Migration-Inducing Protein MIG8. Accession Number: Q9BZZ5. Gene ID: 8539. Expression Region: Pro2~Phe360. Tag Info: N-terminal His Tag. Theoretical MW: 44kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5846