Learn More
Biomatik Corporation Recombinant Colony Stimulating Factor 1, Macrophage (MCSF), 100UG
Supplier:  Biomatik Corporation RPU52501
Encompass_Preferred
Recombinant Mouse Colony Stimulating Factor 1, Macrophage (MCSF) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Colony Stimulating Factor 1, Macrophage (MCSF). Target Synonyms: CSF1; M-CSF; Lanimostim; Processed macrophage colony-stimulating factor 1. Accession Number: P07141. Gene ID: 12977. Expression Region: Lys33~Ser204. Tag Info: N-terminal His Tag. Theoretical MW: 24kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5847