Learn More
Biomatik Corporation Recombinant Asialoglycoprotein Receptor 1 (ASGR1), 100UG
Supplier:  Biomatik Corporation RPU52557
Encompass_Preferred
Recombinant Human Asialoglycoprotein Receptor 1 (ASGR1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Asialoglycoprotein Receptor 1 (ASGR1). Target Synonyms: CLEC4H1; HL-1; ASGPR 1; Asialoglyco protein Receptor; C-type lectin domain family 4 member H1; Hepatic lectin H1. Accession Number: P07306. Gene ID: 432. Expression Region: Phe77~Pro289. Tag Info: N-terminal His Tag. Theoretical MW: 31kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5848