Learn More
Biomatik Corporation Recombinant Ferritin, Light Polypeptide (FTL), 100UG
Supplier:  Biomatik Corporation RPU52655
Encompass_Preferred
Recombinant Mouse Ferritin, Light Polypeptide (FTL) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Ferritin, Light Polypeptide (FTL). Target Synonyms: Ferritin Light Chain; Ferritin Light Polypeptide-Like 3; L Apoferritin; Ferritin L Subunit. Accession Number: P29391. Expression Region: Ile5~Asp183. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5850