Learn More
Biomatik Corporation Recombinant Transmembrane Protein 173 (TMEM173), 100UG
Supplier:  Biomatik Corporation RPU52772
Encompass_Preferred
Recombinant Mouse Transmembrane Protein 173 (TMEM173) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Transmembrane Protein 173 (TMEM173). Target Synonyms: NET23; ERIS; MPYS; MITA; STING; Endoplasmic Reticulum Interferon Stimulator; Stimulator Of Interferon Genes Protein; Mediator of IRF3 activation. Accession Number: Q3TBT3. Gene ID: 72512. Expression Region: Trp160~Leu371. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5852