Learn More
Biomatik Corporation Recombinant Cluster of Differentiation 72 (CD72), 100UG
Supplier:  Biomatik Corporation RPU52821
Encompass_Preferred
Recombinant Mouse Cluster Of Differentiation 72 (CD72) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Cluster Of Differentiation 72 (CD72). Target Synonyms: Lyb2. Accession Number: P21855. Gene ID: 12517. Expression Region: Leu143~Phe345. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5854