Learn More
Biomatik Corporation Recombinant Cancer/Testis Antigen 1B (CTAG1B), 100UG
Catalog No. 501505857
Encompass_Preferred
Recombinant Human Cancer/Testis Antigen 1B (CTAG1B) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cancer/Testis Antigen 1B (CTAG1B). Target Synonyms: CTAG; CTAG1; ESO1; LAGE-2; LAGE2B; NY-ESO-1; CAMEL; ESO2; LAGE-1; LAGE-2b; LAGE2A; Autoimmunogenic cancer/testis antigen NY-ESO-1; L antigen family member 2. Accession Number: P78358. Gene ID: 1485. Expression Region: Met1~Arg180. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5857 Supplier Biomatik Corporation Supplier No. RPU52916
Recombinant Human Cancer/Testis Antigen 1B (CTAG1B) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cancer/Testis Antigen 1B (CTAG1B). Target Synonyms: CTAG; CTAG1; ESO1; LAGE-2; LAGE2B; NY-ESO-1; CAMEL; ESO2; LAGE-1; LAGE-2b; LAGE2A; Autoimmunogenic cancer/testis antigen NY-ESO-1; L antigen family member 2. Accession Number: P78358. Gene ID: 1485. Expression Region: Met1~Arg180. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.