Learn More
Biomatik Corporation Recombinant Actin Beta (ACTb), 100UG
Supplier:  Biomatik Corporation RPU52947
Encompass_Preferred
Recombinant Human Beta Actin (ACTB) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus), Guinea pig (Cavia porcellus), Dog (Canis familiaris; Canine), Pig (Sus scrofa; Porcine), Bovine (Bos taurus), Sheep (Ovis aries), Horse (Equus caballus; Equine), Chicken (Gallus). Target Name: Beta Actin (ACTB). Target Synonyms: Actin Beta; ACT-B; PS1TP5BP1; Actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed. Accession Number: P60709. Gene ID: 60. Expression Region: Met1~Phe375. Tag Info: N-terminal His Tag. Theoretical MW: 44kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5858