Learn More
Biomatik Corporation Recombinant Activin A (ACVA), 100UG
Supplier:  Biomatik Corporation RPU52965
Encompass_Preferred
Recombinant Human Activin A (ACVA) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Activin A (ACVA). Target Synonyms: ACV-A; Activin Beta A Beta A Homodimer. Accession Number: P08476. Gene ID: 3624. Expression Region: Pro44~Gln178. Tag Info: N-terminal His Tag. Theoretical MW: 22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5859