Learn More
Biomatik Corporation Recombinant WAP Four Disulfide Core Domain Protein 2 (WFDC2), 100UG
Supplier:  Biomatik Corporation RPU52977
Encompass_Preferred
Recombinant Human WAP Four Disulfide Core Domain Protein 2 (WFDC2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: WAP Four Disulfide Core Domain Protein 2 (WFDC2). Target Synonyms: WFDC2; HE4; WAP5; EDDM4; Epididymal Protein 4; Epididymal Secretory Protein E4; Putative Protease Inhibitor WAP5; Major epididymis-specific protein E4. Accession Number: Q14508. Gene ID: 10406. Expression Region: Ala30~Phe124. Tag Info: N-terminal His and GST Tag. Theoretical MW: 44kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5861