Learn More
Biomatik Corporation Recombinant Octamer Binding Transcription Factor 4 (OCT4), 100UG
Supplier:  Biomatik Corporation RPU52986
Encompass_Preferred
Recombinant Human Octamer Binding Transcription Factor 4 (OCT4) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Octamer Binding Transcription Factor 4 (OCT4). Target Synonyms: POU5F1; OCT3; OTF3; OTF4; Octamer-4; POU Domain Class 5 Transcription Factor 1; POU Class 5 Homoeobox 1; Octamer-binding protein 3. Accession Number: Q01860. Gene ID: 5460. Expression Region: Ser136~Asn360. Tag Info: N-terminal His Tag. Theoretical MW: 29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5862