Learn More
Biomatik Corporation Recombinant Mannose Associated Serine Protease 1 (MASP1), 100UG
Supplier:  Biomatik Corporation RPU53072
Encompass_Preferred
Recombinant Mouse Mannose Associated Serine Protease 1 (MASP1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Mannose Associated Serine Protease 1 (MASP1). Target Synonyms: CRARF; PRSS5; Mannan-Binding Lectin Serine Peptidase 1; Ra-reactive factor serine protease p100; Complement-activating component of Ra-reactive factor; Serine protease 5. Accession Number: P98064. Gene ID: 17174. Expression Region: His25~Arg453. Tag Info: N-terminal His Tag. Theoretical MW: 53kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5865