Learn More
Biomatik Corporation Recombinant Arginase II (Arg2), 100UG
Supplier:  Biomatik Corporation RPU53145
Encompass_Preferred
Recombinant Mouse Arginase II (Arg2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Arginase II (Arg2). Target Synonyms: Kidney-type arginase; Non-hepatic arginase. Accession Number: O08691. Gene ID: 11847. Expression Region: Val23~Ile354. Tag Info: N-terminal His and GST Tag. Theoretical MW: 66kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5866