Learn More
Biomatik Corporation Recombinant Wingless Type MMTV Integration Site Family, Member 4 (WNT4), 100UG
Supplier:  Biomatik Corporation RPU53287
Encompass_Preferred
Recombinant Human Wingless Type MMTV Integration Site Family, Member 4 (WNT4) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Wingless Type MMTV Integration Site Family, Member 4 (WNT4). Accession Number: P56705. Gene ID: 54361. Expression Region: Leu26~Arg351. Tag Info: N-terminal His Tag. Theoretical MW: 40kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5869