Learn More
Biomatik Corporation BSA Conjugated 17-Hydroxyprogesterone (17-OHP), 100UG
Supplier:  Biomatik Corporation RPU53320
Encompass_Preferred
BSA Conjugated 17-Hydroxyprogesterone (17-OHP) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Universal. Target Name: 17-Hydroxyprogesterone (17-OHP). Target Synonyms: 17OHP; 17-OH Progesterone; 17-Alpha-Hydroxyprogesterone. Tag Info: N-terminal His Tag. Theoretical MW: 330.5Da. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5870