Learn More
Biomatik Corporation Recombinant Fibroblast Growth Factor Receptor 1 (FGFR1), 100UG
Supplier:  Biomatik Corporation RPU53407
Encompass_Preferred
Recombinant Rat Fibroblast Growth Factor Receptor 1 (FGFR1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Fibroblast Growth Factor Receptor 1 (FGFR1). Target Synonyms: CD331; FLT2, KAL2; CEK; FLG; BFGFR, N-SAM,Basic Fibroblast Growth Factor Receptor 1; Fms-Related Tyrosine Kinase-2; Pfeiffer Syndrome. Accession Number: Q04589. Gene ID: 79114. Expression Region: Arg22~Glu376. Tag Info: N-terminal His Tag. Theoretical MW: 48kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5872