Learn More
Biomatik Corporation Recombinant Vimentin (VIM), 100UG
Supplier:  Biomatik Corporation RPU53471
Encompass_Preferred
Recombinant Mouse Vimentin (VIM) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Vimentin (VIM). Accession Number: P20152. Gene ID: 22352. Expression Region: Ser2~Glu466. Tag Info: N-terminal His Tag. Theoretical MW: 52kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5873