Learn More
Biomatik Corporation Recombinant Fibroblast Growth Factor 2, Basic (FGF2), 100UG
Supplier:  Biomatik Corporation RPU53496
Encompass_Preferred
Recombinant Mouse Fibroblast Growth Factor 2, Basic (FGF2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Fibroblast Growth Factor 2, Basic (FGF2). Target Synonyms: B-FGF; BFGF; FGFB; HBGH-2; Basic Fibroblast Growth Factor; Heparin-binding growth factor 2. Accession Number: P15655. Gene ID: 14173. Expression Region: Pro10~Ser154. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5875