Learn More
Biomatik Corporation Recombinant Aquaporin 4 (AQP4), 100UG
Supplier:  Biomatik Corporation RPU53498
Encompass_Preferred
Recombinant Human Aquaporin 4 (AQP4) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Aquaporin 4 (AQP4). Target Synonyms: HMIWC2; MIWC; WCH4; Mercurial-insensitive water channel. Accession Number: P55087. Gene ID: 361. Expression Region: Cys178~Gly317. Tag Info: N-terminal His and GST Tag. Theoretical MW: 45kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5876