Learn More
Biomatik Corporation Recombinant Estrogen Receptor Alpha (ERa), 100UG
Catalog No. 501505878
Encompass_Preferred
Recombinant Human Estrogen Receptor Alpha (ERa) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Estrogen Receptor Alpha (ERa). Target Synonyms: ER-A1; ER; ESR; ESR1; ESRA; NR3A1; NR3-A1; Estrogen Receptor 1; Nuclear Receptor Subfamily 3,Group A,Member 1. Accession Number: P03372. Gene ID: 2099. Expression Region: Ser178~Met438. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5878 Supplier Biomatik Corporation Supplier No. RPU53516
Recombinant Human Estrogen Receptor Alpha (ERa) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Estrogen Receptor Alpha (ERa). Target Synonyms: ER-A1; ER; ESR; ESR1; ESRA; NR3A1; NR3-A1; Estrogen Receptor 1; Nuclear Receptor Subfamily 3,Group A,Member 1. Accession Number: P03372. Gene ID: 2099. Expression Region: Ser178~Met438. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.