Learn More
Biomatik Corporation Recombinant Squamous Cell Carcinoma Antigen 1 (SCCA1), 100UG
Supplier:  Biomatik Corporation RPU53533
Encompass_Preferred
Recombinant Human Serpin B3 (SERPINB3) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Serpin B3 (SERPINB3). Target Synonyms: SCCA1; T4-A; Serpin Peptidase Inhibitor,Clade B(ovalbumin),Member 3; Serpin B3. Accession Number: Q9BYH9. Expression Region: Met1~Ile210. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5879