Learn More
Biomatik Corporation Recombinant S100 Calcium Binding Protein A9 (S100A9), 100UG
Catalog No. 501505880
Encompass_Preferred
Recombinant Bovine S100 Calcium Binding Protein A9 (S100A9) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Bovine (Bos taurus). Target Name: S100 Calcium Binding Protein A9 (S100A9). Target Synonyms: MRP14; MIF; 60B8AG; CFAG; NIF; CAGB; CGLB; L1AG; P14; Calgranulin B; Migration inhibitory factor-related protein 14; Calprotectin L1H subunit; Leukocyte L1 complex heavy chain. Accession Number: P28783. Gene ID: 532569. Expression Region: Ile12~Ala89. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5880 Supplier Biomatik Corporation Supplier No. RPU53554
Recombinant Bovine S100 Calcium Binding Protein A9 (S100A9) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Bovine (Bos taurus). Target Name: S100 Calcium Binding Protein A9 (S100A9). Target Synonyms: MRP14; MIF; 60B8AG; CFAG; NIF; CAGB; CGLB; L1AG; P14; Calgranulin B; Migration inhibitory factor-related protein 14; Calprotectin L1H subunit; Leukocyte L1 complex heavy chain. Accession Number: P28783. Gene ID: 532569. Expression Region: Ile12~Ala89. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.