Learn More
Biomatik Corporation Recombinant Cholinergic Receptor, Nicotinic, Alpha 7 (CHRNa7), 100UG
Supplier:  Biomatik Corporation RPU53584
Encompass_Preferred
Recombinant Human Cholinergic Receptor, Nicotinic, Alpha 7 (CHRNa7) is a purified Prokaryotic Protein. Purity: >80% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cholinergic Receptor, Nicotinic, Alpha 7 (CHRNa7). Target Synonyms: CHR-NA7; CHRN-A7; N-AChRA7; NAChRA7; N-AChR-A7; Neuronal Acetylcholine Receptor Alpha 7. Accession Number: P36544. Gene ID: 1139. Expression Region: Gly23~Thr230. Tag Info: N-terminal His and GST Tag. Theoretical MW: 54kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5881