Learn More
Biomatik Corporation Recombinant Regenerating Islet Derived Protein 1 Beta (REG1b), 100UG
Supplier:  Biomatik Corporation RPU53649
Encompass_Preferred
Recombinant Human Regenerating Islet Derived Protein 1 Beta (REG1b) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Regenerating Islet Derived Protein 1 Beta (REG1b). Target Synonyms: PSPS2; REGH; REGI-BETA; REGL; Pancreatic Stone Protein; Pancreatic Thread Protein; Lithostathine 1 Beta; Secretory Pancreatic Stone Protein 2. Accession Number: P48304. Gene ID: 5968. Expression Region: Gln23~Asn166. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5883