Learn More
Biomatik Corporation Eukaryotic Fibronectin Type III Domain Containing Protein 5 (FNDC5), 100UG
Supplier:  Biomatik Corporation RPU53734
Encompass_Preferred
Eukaryotic Human Fibronectin Type III Domain Containing Protein 5 (FNDC5) is a purified Eukaryotic Protein. Purity: >97% by SDS-PAGE. Host: 293F Cell. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Fibronectin Type III Domain Containing Protein 5 (FNDC5). Target Synonyms: FRCP2; Irisin; Fibronectin type III repeat-containing protein 2. Accession Number: Q8NAU1. Gene ID: 252995. Expression Region: Asp32~Glu143. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5884