Learn More
Biomatik Corporation Recombinant Calumenin (CALU), 100UG
Supplier:  Biomatik Corporation RPU53779
Encompass_Preferred
Recombinant Human Calumenin (CALU) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Calumenin (CALU). Target Synonyms: Crocalbin. Accession Number: O43852. Gene ID: 813. Expression Region: Lys20~Phe315. Tag Info: N-terminal His Tag. Theoretical MW: 49kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5885